Learn More
LATS2, Mouse, Clone: ST-3B11, Abnova™
Mouse monoclonal antibody raised against synthetic phosphopeptide of LATS2.
Marca: Abnova MAB0009.100ug
Descrizione
This gene encodes a serine/threonine protein kinase belonging to the LATS tumor suppressor family. The protein localizes to centrosomes during interphase, and early and late metaphase. It interacts with the centrosomal proteins aurora-A and ajuba and is required for accumulation of gamma-tubulin and spindle formation at the onset of mitosis. It also interacts with a negative regulator of p53 and may function in a positive feedback loop with p53 that responds to cytoskeleton damage. Additionally, it can function as a co-repressor of androgen-responsive gene expression. [provided by RefSeq
Specifica
| LATS2 | |
| Monoclonal | |
| Unconjugated | |
| Western Blot (1-2 ug/mL) ELISA (1 ug/mL) The optimal working dilution should be determined by the end user. | |
| LATS2 | |
| FLJ13161/KPM | |
| Mouse | |
| 100 μg | |
| Primary | |
| Human | |
| Liquid |
| ELISA, Western Blot | |
| ST-3B11 | |
| Mouse monoclonal antibody raised against synthetic phosphopeptide of LATS2. | |
| In 10mM HEPES/KOH, 150mM NaCl, pH 7.5 (50% glycerol) | |
| NM_014572 | |
| LATS2 | |
| Synthetic phosphopeptide (conjugated with KLH) corresponding to residues surrounding S83 of human LATS2. | |
| RUO | |
| 26524 | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG2b |
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.