Learn More
NEFH, Mouse, Clone: RT-97, Abnova™
Mouse monoclonal antibody raised against NEFH.
Marca: Abnova MAB11229.100ug
Descrizione
Neurofilaments are type IV intermediate filament heteropolymers composed of light, medium, and heavy chains. Neurofilaments comprise the axoskeleton and functionally maintain neuronal caliber. They may also play a role in intracellular transport to axons and dendrites. This gene encodes the heavy neurofilament protein. This protein is commonly used as a biomarker of neuronal damage and susceptibility to amyotrophic lateral sclerosis (ALS) has been associated with mutations in this gene. [provided by RefSeq
Specifica
| NEFH | |
| Monoclonal | |
| Unconjugated | |
| Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:200-1:400) The optimal working dilution should be determined by the end user. | |
| NEFH | |
| NEFH | |
| The Triton-X 100 insoluble protein fraction from rat brain. | |
| 100 μg | |
| Primary | |
| Human, Rat | |
| Liquid |
| Immunohistochemistry (PFA fixed) | |
| RT-97 | |
| Mouse monoclonal antibody raised against NEFH. | |
| In PBS (0.05% BSA, 0.05% sodium azide) | |
| NFH | |
| Mouse | |
| Protein G affinity chromatography | |
| RUO | |
| 4744 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG1 κ |
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.