Learn More
Mouse anti-Rat IgA Heavy Chain Secondary Antibody (MARA-1), DyLight 550, Novus Biologicals™
Mouse Monoclonal Antibody
Marca: Novus Biologicals NBP2-68392R
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
IgA Heavy Chain Monoclonal antibody specifically detects IgA Heavy Chain in Rat samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry (Frozen), Immunofluorescence
Specifica
| IgA Heavy Chain | |
| Monoclonal | |
| DyLight 550 | |
| Mouse | |
| Affinity purified | |
| RUO | |
| Rat | |
| Purified |
| ELISA, Immunohistochemistry, Immunohistochemistry (Frozen), Immunofluorescence | |
| MARA-1 | |
| 50mM Sodium Borate | |
| Purified IR1060 IgA rat myeloma protein | |
| 0.25 mL | |
| Secondary | |
| Store at 4°C in the dark. | |
| IgG1 |