Learn More
Invitrogen™ Mouse anti-Rat Kappa Light Chain Secondary Antibody, FITC
Mouse Monoclonal Secondary Antibody
Marca: Invitrogen™ SA510180
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
Mouse anti-Rat Kappa Light Chain Secondary Antibody for Flow
Anti-Rat secondary antibodies are affinity-purified antibodies with well-characterized specificity for rat immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
Specifica
| Rat Kappa Light Chain | |
| Monoclonal | |
| 0.1 mg/mL | |
| PBS with 4-5mg/mL BSA and 0.02% sodium azide | |
| Pooled rat serum IgG. | |
| 100 μg | |
| Secondary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. Store in the dark. | |
| Liquid |
| Flow Cytometry | |
| OX-12 | |
| FITC | |
| Mouse | |
| Protein G | |
| RUO | |
| Rat | |
| Secondary Antibody | |
| IgG2a |