missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse C-Reactive Protein/CRP Antibody, R&D Systems™

Rat Monoclonal Antibody
Marca: R&D Systems MAB1829-SP
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
C-Reactive Protein/CRP Monoclonal specifically detects C-Reactive Protein/CRP in Mouse samples. It is validated for ELISA Capture (Matched Antibody Pair).
Specifica
| C-Reactive Protein/CRP | |
| Monoclonal | |
| LYOPH | |
| ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
| P14847 | |
| CRP | |
| Mouse myeloma cell line NS0-derived recombinant mouse C-Reactive Protein Met1-Ser225 Accession # P14847 | |
| 25 μg | |
| Primary | |
| Detects mouse C-Reactive Protein in ELISAs. | |
| Mouse | |
| Purified |
| ELISA | |
| 267917 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| C-reactive protein, C-reactive protein, pentraxin-related, CRP, MGC88244, pentraxin 1, PTX1MGC149895 | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 1401 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
For Research Use Only
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto