missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse CCL17/TARC Antibody, R&D Systems™

Rat Monoclonal Antibody has been used in 15 publications
Marca: R&D Systems MAB529-500
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
CCL17/TARC Monoclonal specifically detects CCL17/TARC in Mouse samples. It is validated for Neutralization, ELISA Capture (Matched Antibody Pair).
Specifica
| CCL17/TARC | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| ABCD-2, CC chemokine TARC, C-C motif chemokine 17, chemokine (C-C motif) ligand 17, MGC138273, SCYA17, SCYA17MGC138271, small inducible cytokine subfamily A (Cys-Cys), member 17, Small-inducible cytokine A17, T cell-directed CC chemokine, TARC, TARCA-152E5.3, Thymus and activation-regulated chemokine | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 6361 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
| Neutralization, ELISA | |
| 110904 | |
| Neutralization 0.75-3.0 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
| Q9WUZ6 | |
| CCL17 | |
| E. coli-derived Recombinant Mouse CCL17/TARC Ala24-Pro93 Accession # Q9WUZ6 | |
| 500 μg | |
| Primary | |
| Detects mouse CCL17/TARC in ELISAs. | |
| Mouse | |
| Purified |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
For Research Use Only
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto