Learn More
ACROBiosystems Mouse CEACAM-5 Protein, His Tag
Marca:  ACROBiosystems CE5-M52H3-1mg
Dettagli aggiuntivi : Peso : 0.00010kg
Codice prodotto. 17276170
  • 2874.14€ / 1mg
Spedizione stimata: 07-08-2024
per visualizzare lo stock disponibile
Descrizione
Descrizione
Carcinoembryonic antigen-related cell adhesion molecule 5 (CEACAM5) is also Carcinoembryonic antigen (CEA), Meconium antigen 100, CD antigen CD66e, CEACAM5 belongs to the immunoglobulin superfamily and CEA family. CEACAM5 contains seven Ig-like (immunoglobulin-like) domains. CEACAM5 is homodimer protein which binding of E.coli Dr adhesins leads to dissociation of the homodimer. CEACAM5 is cell surface glycoprotein that plays a role in cell adhesion and in intracellular signaling. CEACAM5 is receptor for E.coli Dr adhesins.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 104.5 kDa.
Specifica
Specifica
104.5 kDa
Mouse CEACAM-5, His Tag (CE5-M52H3) is expressed from human 293 cells (HEK293). It contains AA Gln 35 - Glu 947 (Accession # Q3UKK2-1).
-20°C
CEACAM-5
Mouse
HEK293
Bio-Markers & CD Antigens
1 mg
Gln 35 - Glu 947
CEACAM-5,CD66e,CEA,Meconium antigen 100
Unconjugated
His Tag
Powder