missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Mouse HB-EGF (NP_034545) VersaClone cDNA
Click to view available options
Quantità:
10 μg
Dimensione della confezione:
10µg
Descrizione
This shuttle vector contains the complete ORF for the gene of interest, along with a Kozak consensus sequence for optimal translation initiation. It is inserted NotI to AscI. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector.
The Mouse HB-EGF (NP_034545) VersaClone cDNA clone contains the complete ORF for HB-EGF, along with a Kozak consensus sequence for optimal translation initiation. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector. The HB-EGF insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.
Order Info
Ships at ambient temperature
Specifica
Specifica
| Gene | HBEGF |
| Numero di accesso | NP_034545 |
| Concentrazione | 10 μg at 0.2 μg/μL |
| Formulazione | cDNA is provided in 10mM Tris-Cl, pH 8.178 |
| Tipo di inserto | 640bp |
| ID gene (immissione) | 1839 |
| Alias gene | diphtheria toxin receptor (heparin-binding epidermal growth factor-like growthfactor), Dtr, DTRHEGFLdiphtheria toxin receptor (heparin-binding EGF-like growth factor), Dts, DTSF, HBEGF, Hegfl, heparin-binding EGF-like growth factor, heparin-binding epidermal growth factor, proheparin-binding EGF-like growth factor |
| Content And Storage | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Specie | Mouse |
| Quantità | 10 μg |
For Reasearch Use Only. Not for use in Humans.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction