missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse IgG2A Antibody, R&D Systems™

Rat Monoclonal Antibody
Marca: R&D Systems MAB0032-100
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
IgG2a Monoclonal specifically detects IgG2a in Mouse samples. It is validated for Flow Cytometry.
Specifica
| IgG2a | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. | |
| Cocktail of mouse IgG 1 and IgG 2A | |
| 100 μg | |
| Secondary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
| Flow Cytometry | |
| 344701 | |
| Flow Cytometry 0.25 ug/10^6 cells | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| The specificity of this antibody was confirmed by direct ELISA on mouse IgG 2A monoclonal antibodies, and it does not cross-react with IgG 1 IgG 2B IgG 3 IgM, IgA, or IgE antibodies. By flow cytometry, this isotype-specific secondary antibody detects primary antibodies of the mouse IgG 2A isotype, and does not detect primary antibodies of mouse IgG 1 IgG 2B or IgG 3 class. | |
| Mouse | |
| Purified |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
RUO
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto