missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Mouse LILRB4/CD85k/ILT3 (NP_038560) VersaClone cDNA
Click to view available options
Quantità:
10 μg
Dimensione della confezione:
10µg
Descrizione
This shuttle vector contains the complete ORF for the gene of interest, along with a Kozak consensus sequence for optimal translation initiation. It is inserted NotI to AscI. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector.
The Mouse LILRB4/CD85k/ILT3 (NP_038560) VersaClone cDNA clone contains the complete ORF for ILT3/CD85k/LILRB4, along with a Kozak consensus sequence for optimal translation initiation. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector. The ILT3/CD85k/LILRB4 insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.
Order Info
Ships at ambient temperature
Specifica
Specifica
| Gene | ILT3/CD85k/LILRB4 |
| Numero di accesso | NP_038560 |
| Concentrazione | 10 μg at 0.2 μg/μL |
| Formulazione | cDNA is provided in 10mM Tris-Cl, pH 8.98 |
| Tipo di inserto | 1021bp |
| ID gene (immissione) | 11006 |
| Alias gene | CD85 antigen-like family member K, CD85k, CD85k antigen, HM18, ILT3, ILT-3, ILT3CD85K, leukocyte immunoglobulin-like receptor, subfamily B (with TM and ITIM domains), LIR5, LIR5LILRB5, LIR-5 subfamily B, member 4, member 4 |
| Content And Storage | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Specie | Mouse |
| Quantità | 10 μg |
For Reasearch Use Only. Not for use in Humans.
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto