Learn More
R&D Systems™ Mouse MS4A8A (AAK37415) VersaClone cDNA
Click to view available options
Quantità:
10 μg
Dimensione della confezione:
10µg
Descrizione
The Mouse MS4A8A (AAK37415) VersaClone cDNA clone contains the complete ORF for MS4A8A, along with a Kozak consensus sequence for optimal translation initiation. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector. The MS4A8A insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.
Order Info
Ships at ambient temperature.
Specifica
Specifica
| Alias gene | 2010004L09Rik, AI481240, CD20 antigen-like 5, CD20L, CD20L5, membrane-spanning 4-domains subfamily A member 8, Ms4, MS4A8, MS4a8B protein |
| Content And Storage | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Quantità | 10 μg |
For Reasearch Use Only; Not for Use in Humans