missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Mouse OX40/TNFRSF4 (NP_035789) VersaClone cDNA
Click to view available options
Quantità:
10 μg
Dimensione della confezione:
10µg
Descrizione
The Mouse OX40/TNFRSF4 (NP_035789) VersaClone cDNA clone contains the complete ORF for OX40/TNFRSF4, along with a Kozak consensus sequence for optimal translation initiation. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector. The OX40/TNFRSF4 insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.
Order Info
Ships at ambient temperature.
Specifica
Specifica
| Gene | OX40/TNFRSF4 |
| Numero di accesso | NP_035789 |
| ID gene (immissione) | 7293 |
| Alias gene | ACT-135, ACT35 antigen, ACT35ATC35 antigen, CD134, CD134 antigen, Ly-70, O x 40 cell surface antigen, O x 40 homologue, O x 40 L receptor, O x 40 lymphoid activation antigene ACT35, TAX transcriptionally-activated glycoprotein 1 receptor, tax-transcriptionally activated glycoprotein 1 receptor, TNFRSF4, tumor necrosis factor receptor superfamily member 4, tumor necrosis factor receptor superfamily, member 4, Txgp1, TXGP1L, TXGP1LO x 40 antigen |
| Content And Storage | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Specie | Mouse |
| Quantità | 10 μg |
For Reasearch Use Only; Not for Use in Humans
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto