missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse/Porcine 5'-Nucleotidase/CD73 Antibody, R&D Systems™
Sheep Polyclonal Antibody has been used in 2 publications
Marca: R&D Systems AF4488
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
5'-Nucleotidase/CD73 Polyclonal specifically detects 5'-Nucleotidase/CD73 in Mouse, Porcine samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, CyTOF-ready.
Specifica
| 5'-Nucleotidase/CD73 | |
| Polyclonal | |
| Unconjugated | |
| Q61503 | |
| NT5E | |
| Chinese hamster ovary cell line CHO-derived recombinant mouse 5'-Nucleotidase/CD73 Trp29-Lys549 Accession # Q61503 | |
| 100 μg | |
| Primary | |
| Detects mouse 5'-Nucleotidase/CD73 in direct ELISAs and Western blots. | |
| Mouse, Pig | |
| IgG |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, CyTOF | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, Immunohistochemistry 5-15 ug/mL, Immunocytochemistry 5-15 ug/mL, CyTOF-ready | |
| 5' nucleotidase (CD73), 5'-NT, 5-NT, 5'-nucleotidase, ecto (CD73), CD73, CD73 antigen, E5NT, EC 3.1.3.5, ecto-5'-nucleotidase, eN, eNT, NT, NT55'-nucleotidase, NT5E, NTE, Purine 5-Prime-Nucleotidase | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 4907 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto