missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse TREM2 Antibody, R&D Systems™

Rat Monoclonal Antibody has been used in 5 publications
Marca: R&D Systems MAB1729-SP
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
TREM2 Monoclonal specifically detects TREM2 in Mouse samples. It is validated for Western Blot.
Specifica
| TREM2 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| Q99NH8 | |
| TREM2 | |
| Mouse myeloma cell line NS0-derived recombinant mouse TREM-2 Leu19-Pro168 Accession # Q99NH8 | |
| 25 μg | |
| Primary | |
| Detects mouse TREM-2 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant mouse (rm) TREM-1, recombinant human TREM-2, or rmTREM-3 is observed. | |
| Mouse | |
| Purified |
| Western Blot | |
| 237916 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| PLOSL2, TREM-2, Trem2a, Trem2b, Trem2c, TREM-2triggering receptor expressed on myeloid cells 2a, Triggering receptor expressed on monocytes 2, triggering receptor expressed on myeloid cells 2 | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 54209 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
For Research Use Only
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto