Learn More
MP Biomedicals™ Tryptic Soy Broth
Sfoglia Tutto MP Biomedicals Prodotti
Click to view available options
:
Tryptic Soy Broth; 500g
Specifica
Specifica
| Content And Storage | 15°C to 30°C |
| Forma | Powder |
| Tipo di media | Tryptic Soy Broth |
| Product Type | Microbiology Media |
| Formulazione | Casein digest Peptone 17g/L, Soy Peptone 3g/L, Disodium Phosphate 2.5g/L, Dextrose 2.5g/L, Sodium Chloride 5g/L |
| Da utilizzare con (applicazione) | Isolation and cultivation of non-fastidious organisms |
| Quantità | 500 g |