Learn More
MP Biomedicals™ X-Gal Powder
Codice prodotto. 11317849 Sfoglia Tutto MP Biomedicals Prodotti
Click to view available options
Quantità:
1 g
Dimensione della confezione:
1g
Codice prodotto. 11317849
Marca: MP Biomedicals™ 114063102
Used to detect β-galactosidase activity to differentiate recombinants from nonrecombinants in cloning experiments
Protein trafficking; Cell viability; Cytotoxicity;Gene expression; Transformation; Cloning and expression; Bacteria cell culture; Microbiological analysis
TRUSTED_SUSTAINABILITY
Specifica
Content And Storage 15° to 30°C
Quantità 1 g