missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MTM1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 13924-1-AP-150UL
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
Human MTM1, a 603 amino-acid protein, is mutated in myotubular myopathy. The largely related protein hMTMR2 is found mutated in a recessive form of Charcot-Marie-tooth neuropathy. Myotubularin is primarily a lipid phosphatase that acts on hosphatidylinositol 3-monophosphate and is involved in the regulation of the phosphatidylinositol 3-kinase (PI 3-kinase) pathway and membrane trafficking. Wildtype myotubularin can directly dephosphorylate PI 3-P and PI 4-P in vitro. Thus, it decreases PI 3-P levels by down-regulating PI 3-K activity and by facilitating the degradation of PI 3-P.Specifica
| MTM1 | |
| Polyclonal | |
| Unconjugated | |
| MTM1 | |
| CG2, CNM, MTM1, MTMX, Myotubularin, myotubularin 1, XLMTM | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 17772, 288762, 4534 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| 0.35 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q13496, Q6AXQ4, Q9Z2C5 | |
| Mtm1 | |
| MTM1 Fusion Protein Ag4940 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto