Learn More
Mubritinib, MedChemExpress
Product Code. 16441877
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16441877 50 mg 50mg
16425718 10 mg 10mg
16436206 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 16441877 Supplier Medchem Express Supplier No. HY13501
 Restricted Item
Spedizione stimata: 30-11-2026
MedChemExpress Mubritinib (TAK-165) is a potent and selective EGFR2/HER2 inhibitor with an IC50 of 6 nM.
Mubritinib (TAK-165) is a potent and selective EGFR2/HER2 inhibitor with an IC50 of 6 nM.
Specifications
Chemical Name or Material Mubritinib
CAS 366017-09-6
Color White
Purity Grade Notes Research
Molecular Formula C25H23F3N4O2
Quantity 50 mg
Synonym TAK-165
Solubility Information DMSO : 50 mg/mL (106.73 mM; Need ultrasonic)
SMILES FC(C1=CC=C(/C=C/C2=NC(COC3=CC=C(CCCCN4N=NC=C4)C=C3)=CO2)C=C1)(F)F
Molecular Weight (g/mol) 468.47
Formula Weight 468.47
Percent Purity 99.85%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less
Research purposes only