Learn More
MUC2 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 27675-1-AP-150UL
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
Secreted epithelial mucins are large macromolecules which exhibit extreme polydispersity. Mucin 2 is the major intestinal mucin. O-glycans are attached to MUC2 in a potentially diverse arrangement, which is crucial for their interaction with endogeneous and exogeneous lectins.Specifica
| MUC2 | |
| Polyclonal | |
| Unconjugated | |
| MUC2 | |
| Intestinal mucin 2, MLP, MUC 2, MUC2, Mucin 2, SMUC | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 17831, 4583 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.43 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q02817, Q80Z19 | |
| Muc2 | |
| MUC2 Fusion Protein Ag25800 | |
| 150 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |