Learn More
Heidolph™ Pompa a testa multi-canale
Sfoglia Tutto Heidolph Instruments Prodotti
Click to view available options
Modello:
C 12
C 4
C 8
Capacità:
12 small cassettes
4 small cassettes
8 medium cassettes
Include:
4 rollers
8 rollers
Dimensione della confezione:
Pezzo
Descrizione
- Separate metering into multiple vessels with different feed rates in a single operation by using different tubing dimensions to adjust flow rate
- Save time - tubing change is done in just seconds
- Change your cassettes easily, even during operation – there are no restrictions
- Cassette adjustments and changes are simple, just click in place
- No additional purchase of a pump drive needed: upgrade your single-channel pump drive with just one adaptor for multi-channel use and pick the head/cassette configuration that matches your application needs
Specifica
Specifica
| Capacità | 4 small cassettes |
| Include | 8 rollers |
| Modello | C 4 |
| Tipo | Pump Head |