Learn More
Multi-Element Organometallic Oil Standards 5 Element, SPEX CertiPrep™
Sfoglia Tutto SPEX Certiprep Prodotti
Click to view available options
Quantità:
100 g
Dimensione della confezione:
100g
Descrizione
Includes
NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
| Analyte | Concentration |
|---|---|
| Ba | 5000μg/g |
| Ca | 5000μg/g |
| Mg | 5000μg/g |
| P | 5000μg/g |
| Zn | 5000μg/g |
Certifications
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Compliance
GHS/OSHA Complaint
Specifica
Specifica
| Materiale o nome chimico | 5 Element Metal Additive Oil Standard |
| Imballaggio | Triple-leached LDPE bottle |
| Concentration or Composition Notes | Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis. |
| Nomi componenti o analita | Ba,Ca,Mg |
| Da utilizzare con (apparecchiatura) | AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation |
| Tipo | Certified Reference Material |
| Elemento | Ba, Ca, Mg |
| Solvent or Matrix | Base Oil 75 |
| Industry Type | Petrochemical Testing |
| Quantità | 100 g |
Product intended for laboratory use only