Learn More
Multi-Parameter Refractive Index & Density Certified Reference Material, at 15 °C, 20 °C and 25 °C, UKAS ISO 17025 / ISO 17034 Certified, Single 30 mL (Nominal RI Value 1.6579 at 20 °C), Paragon Scientific™
Codice prodotto. 16080272
Click to view available options
Quantità:
30 mL
Dimensione della confezione:
30mL
Codice prodotto. 16080272
Marca: Paragon Scientific PSRVD04
Manufactured for refractive index and density measurements. The combined Refractive Index and Density CRM is available in four different materials, with each material including certified data for both refractive index and density at 15 °C, 20 °C and 25 °C.
  • Multi-Parameter Certified Reference Materials for Refractive Index and Density at 15 °C, 20 °C and 25 °C
  • Certified in strict accordance to ISO 17025 and ISO 17034 under our UKAS accreditation
  • Highest level of accreditation guarantee, providing the most credible certified data available worldwide
  • Density certified in accordance with primary level ASTM D1480 methodology
  • Ensures full compliance to ASTM and IP test method protocols
  • All measurements are fully traceable to NIST and international protocols
  • Low levels of uncertainty, ensuring maximum accuracy of data and dependable results
  • Supplied in 30 mL volume sealed glass vials for ample measurement readings
  • 12 month shelf life
Specifica
Quantità 30 mL
compliance-icons
Identificativo prodotto
  • 30ML Multi-Parameter Refractive Index & Density St andard
Avvertenza
  • Attenzione
Categoria di pericolo
  • Tossicità acuta Categoria 4
  • Lesione oculare grave/irritazione oculare Categoria 2
Indicazioni di pericolo
  • H302-Nocivo se ingerito.
  • H319-Provoca grave irritazione oculare.
Consigli di prudenza
  • P301+P312-IN CASO DI INGESTIONE accompagnata da malessere: contattare un CENTRO ANTIVELENI/medico/
  • P305+P351+P338-IN CASO DI CONTATTO CON GLI OCCHI: sciacquare accuratamente per parecchi minuti. Togliere le eventuali lenti a contatto se è agevole farlo. Continuare a sciacquare.
Informazioni supplementari
  • MIXTURE LIST-Contiene: 1-Bromonaphthalene
Nominal values quoted. Certified Values will vary from batch to batch.