Learn More
MP Biomedicals™ Multitest Slide
Sfoglia Tutto MP Biomedicals Prodotti
Click to view available options
Lunghezza (sistema metrico):
2 mm
3 mm
Larghezza (sistema metrico):
4 mm
5 mm
6 mm
Diametro (sistema metrico):
4 mm
6 mm
7 mm
8 mm
Dimensione della confezione:
pezzi 100
Descrizione
The uncoated areas have a total wettable surface so that liquid spreads evenly over the area of each well. For cell attachment and growth, the slides require only a simple washing in deionized water or detergent followed by sterilization.
- Self-draining surface
- Sterilizable
- Resistant to direct heat
- Resistant to common organic solvents and fixatives
- One-time use
- Numbered
Specifica
Specifica
| Lunghezza (sistema metrico) | 3 mm |
| Larghezza (sistema metrico) | 5 mm |
| Tipo | Cell Culture Slide |
| Conteggio pozzetti | 15 |
| Diametro (sistema metrico) | 4 mm |
| Quantità | 100/Box |
Unless specified otherwise, for research or further manufacturing use only, not for direct human use.