missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ P110 (mitochondrial membrane protein) Monoclonal Antibody (2G2)

Mouse Monoclonal Antibody
Marca: Invitrogen™ MA124690
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
MA1-24690 detects Mitochondrial p110 from human and monkey samples.
P110 is a human mitochondrial protein with a molecular weight of 110 kDa. Close examination of the staining pattern in HeLa and Fanconi's Anemia cells reveal differences in the morphology and organization of mitochondria in these two cell types. The epitope targeted may serve as a valuable marker in the investigation of relationships between mitochondria and other cellular structures in human cells. The mouse monoclonal P110 antibody labels mitochondria in human cells, as assessed by double staining with either Rhodamine 123 or a polyclonal antibody to mitochondrial matrix HSP-60 proteins. The P110 antigen has an approximate isoelectric point of 6.5 that copartitions with HSP-60 proteins during isolation of mitochondria from HeLa cells. The P110 staining pattern in HeLa and Fanconi's anaemia cells reveals differences in the morphology and organization of mitochondria in these two cell types. In HeLa cells, mitochondria appear as individual tubular compartments of variable length and are closely associated with vimentin filaments, particularly at the periphery of the nucleus. In Fanconi's anaemia cells, mitochondria have a filamentous shape and form an interconnected cytoplasmic reticulum running in parallel with both vimentin filaments and microtubules. After stabilization with aldehyde- or alcohol-based fixation protocols that optimize the preservation of cytoskeletal components, the epitope targeted by the 2G2 antibody may serve as a valuable marker in the investigation of relationships between mitochondria and other cellular structures in human cells.
Specifica
| P110 (mitochondrial membrane protein) | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 0.08% sodium azide; pH 7.4 | |
| Q14764 | |
| MVP | |
| Recombinant full length human mitochondrial p110 protein. | |
| 25 μg | |
| Primary | |
| Human, Monkey | |
| Antibody | |
| IgG1 |
| Western Blot | |
| 2G2 | |
| Unconjugated | |
| MVP | |
| 2310009M24Rik; LRP; Lung resistance-related protein; major vault protein; MVP; MVP1; Testicular Secretory Protein Li 30; VAULT1; vRNP | |
| Mouse | |
| Protein G | |
| RUO | |
| 9961 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto