Learn More
Invitrogen™ MYH11 Polyclonal Antibody

Rabbit Polyclonal Antibody
Marca: Invitrogen™ PA5116524
Descrizione
Positive Samples: LO2, A375, Mouse liver, Mouse brain, Rat liver Immunogen sequence: ATAKENEKKA KSLEADLMQL QEDLAAAERA RKQADLEKEE LAEELASSLS GRNALQDEKR RLEARIAQLE EELEEEQGNM EAMSDRVRKA TQQAEQLSNE LATERSTAQK NESARQQLER QNKELRSKLH EMEGAVKSKF KSTIAALEAK IAQLEEQVEQ EAREKQAATK SLKQKDKKLK EILLQVEDER KMAEQYKEQA EKGNARVKQL KRQLEEAEEE SQRINANRRK LQRELDEATE SNEAMGREVN ALKSKLRRGN ETSFVPSRRS GGRRVIENAD GSEEETDTRD ADFNGTKASE.
SM-MHC is a cytoplasmic structural protein, which is a major component of the contractile apparatus in smooth muscle cells. Expression of smooth muscle myosin is developmently regulated, appearing early in smooth muscle development, and is specific for smooth muscle development. Two isoforms of smooth muscle myosin heavy chain have been identified, designated MHC-1 and MHC-2.
Specifica
| MYH11 | |
| Polyclonal | |
| Unconjugated | |
| MYH11 | |
| AAT4; AV071570; contractile protein; FAA4; KIAA0866; LOW QUALITY PROTEIN: myosin-11; Myh11; myh11 {ECO:0000312; Myosin heavy chain 11; myosin heavy chain 11, smooth muscle; myosin heavy chain 21; myosin heavy chain isoform SMA; myosin heavy chain SMA; myosin heavy chain, smooth muscle isoform; myosin, heavy chain 11, smooth muscle; myosin, heavy polypeptide 11, smooth muscle; myosin-11; RGD:3136}; SM1; SM2; SMHC; SMMHC; smooth muscle myosin heavy chain | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 17880, 24582, 4629 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot, Immunocytochemistry | |
| 0.37 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3 | |
| O08638, P35749, Q63862 | |
| MYH11 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1680-1979 of human SMMHC/SMMHC/MYH11 (NP_0010352031). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.