Learn More
N-Boc-O-tert-butyl-L-threonine, 95%
Codice prodotto. p-7054479
Click to view available options
Quantità:
1 g
5 g
Dimensione della confezione:
1g
5g
Codice prodotto. 15459198
Marca: Thermo Scientific Alfa Aesar H62713.03
Richiesta di formato massivo o personalizzato
CAS: 13734-40-2 | C13H25NO5 | 275.35 g/mol
N-Boc-O-tert-butyl-L-threonine is used as pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
N-Boc-O-tert-butyl-L-threonine is used as pharmaceutical intermediate.
Solubility
Soluble in DMF.
Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY
Identificatori chimici
CAS 13734-40-2
Formula molecolare C13H25NO5
Molecular Weight (g/mol) 275.35
Numero MDL MFCD00151115
InChI Key LKRXXARJBFBMCE-BDAKNGLRSA-N
Sinonimo boc-thr tbu-oh, boc-o-tert-butyl-l-threonine, boc-o-t-butyl-l-threonine, 2s,3r-3-tert-butoxy-2-tert-butoxycarbonyl amino butanoic acid, l-threonine, n-1,1-dimethylethoxy carbonyl-o-1,1-dimethylethyl, boc-thr t-bu-oh, pubchem12289, boc-l-thr tbu-oh, boc-o-tert-butyl-l-thr-oh
PubChem CID 7015770
IUPAC Name (2S,3R)-3-[(2-methylpropan-2-yl)oxy]-2-[(2-methylpropan-2-yl)oxycarbonylamino]butanoic acid
SMILES C[C@@H](OC(C)(C)C)[C@H](NC(=O)OC(C)(C)C)C(O)=O
Specifica
Materiale o nome chimico N-Boc-O-tert-butyl-L-threonine
Punto di fusione 95°C to 98°C
Beilstein 4454820
Informazioni di solubilità Soluble in DMF.
Quantità 1 g
Peso formulazione 275.34
Percent Purity 95%
Forma fisica Solid