Learn More
N,N-Dimethylsuccinamic acid, 98%
Codice prodotto. p-7044937
Click to view available options
Quantità:
5 g
25 g
Dimensione della confezione:
25g
5g
Codice prodotto. 11322716
Marca: Thermo Scientific Alfa Aesar L08796.06
Richiesta di formato massivo o personalizzato
CAS: 2564-95-6 | C6H11NO3 | 145.158 g/mol
It is employed as a intermediate for pharmaceutical.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is employed as a intermediate for pharmaceutical.
Solubility
Slightly soluble in water.
Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY
Identificatori chimici
CAS 2564-95-6
Formula molecolare C6H11NO3
Molecular Weight (g/mol) 145.158
Numero MDL MFCD00043505
InChI Key WAIGPJMZARQZDX-UHFFFAOYSA-N
Sinonimo n,n-dimethylsuccinamic acid, 4-dimethylamino-4-oxobutanoic acid, nn-dimethylsuccinamic acid, butanoic acid, 4-dimethylamino-4-oxo, n,n-dimethylsuccinic acid monoamide, 3-dimethylcarbamoyl propanoic acid, succinamic acid, n,n-dimethyl, n-dimethyl-succinamic acid, acmc-20ak89, succinamic acid,n-dimethyl
PubChem CID 75717
IUPAC Name 4-(dimethylamino)-4-oxobutanoic acid
SMILES CN(C)C(=O)CCC(=O)O
Specifica
Materiale o nome chimico N,N-Dimethylsuccinamic acid
Punto di fusione 82°C to 87°C
Quantità 5 g
Beilstein 1762788
Informazioni di solubilità Slightly soluble in water.
Peso formulazione 145.16
Percent Purity 98%