Learn More
Thermo Scientific™ Nalgene™ Boston Round Narrow-Mouth Natural PPCO Bottles with Closure: Bulk Pack
GREENER_CHOICE
Codice prodotto. 10622064
Click to view available options
Capacità (sistema metrico):
30 mL
60 mL
125 ml
250 mL
500 mL
1 L
Dimensione chiusura:
20 mm
24 mm
28 mm
38-430 mm
Dimensione della confezione:
pezzi 1000
pezzi 125
pezzi 250
pezzi 50
pezzi 500
Codice prodotto. 10622064
Marca: Thermo Scientific™ 3120060016
Solo null disponibili
Aggiungi al carrello
Aggiungi al carrello
Ensure continued stock of these narrow-mouth natural PPCO bottles with closure with this bulk-packed option.
Ensure continued stock of translucent, durable and chemically resistant Thermo Scientific™ Boston Round Narrow-Mouth Natural PPCO Bottles with Closure with this bulk-packed option. These bottles are constructed from natural polypropylene copolymer and feature polypropylene closures.
Features:
  • Durable
  • Translucent
  • Autoclavable/Leakproof†
TRUSTED_SUSTAINABILITY
Specifica
Capacità 16 oz., 500 mL
Capacità (sistema metrico) 500 mL
Capacità (sistema imperiale) 16 oz.
Colore Natural
Descrizione Narrow-Mouth Boston Round Bottle
Diametro (sistema metrico) interno collo 21 mm
Materiale PPCO
N. per custodia 125
N. per confezione 125
Sterilità Non-sterile
Colore chiusura Natural
Finitura collo 28 mm
Certificazione/Conformità Manufactured in compliance with ISO 13485:2016 quality requirements. Bottles and Closures meet the requirements of 21CFR177.1520 for food and beverage use, USP Class VI.
Closure Included Yes
Graduated No
Bocca Narrow
Forma Round
Altezza (sistema metrico) 170 mm
Product Type Narrow Mouth Bottle
Larghezza (sistema imperiale) 2.86 in.
Altezza (sistema imperiale) 6.71 in.
Rivestimento interno chiusura Unlined
Materiale chiusura Polypropylene
Dimensione chiusura 28 mm
Diametro (sistema metrico) 73 mm
Tipo di chiusura Yes (Bulk Packed, Unassembled)
Erogazione attraverso la chiusura No
Autoclavabile Autoclavable
† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for two minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.