Learn More
Thermo Scientific™ Nalgene™ Heavy-Duty Cylindrical LLDPE Tanks with Spigots
Codice prodotto. 10384541
Click to view available options
Capacità (sistema metrico):
19 L
26.5 L
37.8 L
56.8 L
113.5 L
208 L
Dimensione della confezione:
Pezzo
Codice prodotto. 10384541
Marca: Thermo Scientific™ 111020007
Solo null disponibili
Aggiungi al carrello
Aggiungi al carrello
Nalgene Heavy-Duty Cylindrical LLDPE Tanks include a needle-type spigot installed for draw off. Spigot accepts 5/8 inch I.D. tubing. Tanks are offered in sizes from 19 to 208L.
Thermo Scientific™ Nalgene Heavy-Duty Cylindrical LLDPE Tanks include a needle-type spigot installed for draw off. This spigot accepts 5/8 inch I.D. tubing and are available in sizes ranging from 19L to 208L.
  • Needle Type Tank Spigot Nalgene (Cat. No. 6421) is installed in tank to allow draw off
  • 5/8 in. (15.9mm) I.D. tubing
  • Offers good temperature and chemical resistance
  • Not autoclavable
Includes:
Tank cover included
Note:
Tanks used at elevated temperatures or with high specific-gravity liquids should be externally supported.
TRUSTED_SUSTAINABILITY
Specifica
Diametro (sistema imperiale) esterno 12 in.
Diametro (sistema metrico) esterno 32 cm
Tipo Tank with Spigot
Materiale LLDPE
Capacità (sistema metrico) 26.5 L
Altezza (sistema imperiale) 18 in.
Altezza (sistema metrico) 46 cm
Disclaimer to internal research use restriction:
Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Caution:
Plastic tanks are generally subject to more severe conditions that plastic labware; exposure is constant, stresses are greater and different classes and concentrations of chemicals are involved. Please pay special attention to chemical compatibility.