Learn More
Thermo Scientific™ Nalgene™ LDPE Low Particulate/Low Metals Bottles with Closure

Descrizione
Accurately pour and fill Thermo Scientific™ Nalgene™ LDPE Low Particulate/Low Metals Bottles with Closure, which feature a narrow mouth. These low-density polyethylene bottles include certification to contain less than 20 particles per mL at 0.3 microns or greater.
- Certified to particulate and trace metals levels
- Linerless polypropylene closures provide the ultimate in leakproof protection without the use of liner that can wrinkle, cause leaks or contaminate your reagents
- Exhibits flexibility, impact resistance and excellent visibility of contents for a wide variety of uses in the lab
- Excellent chemical resistance to most acids, bases and alcohols make this a versatile bottle
- Translucent
- Autoclavable/Nonsterile
- Guaranteed leakproof†
Specifica
Specifica
| Capacità | 8 oz., 250 mL |
| Capacità (sistema metrico) | 250 mL |
| Capacità (sistema imperiale) | 8 oz. |
| Colore | Translucent |
| Descrizione | Low Particulate/Low Metals Bottle |
| Diametro (sistema metrico) interno collo | 18 mm |
| Materiale | LDPE |
| N. per custodia | 72 |
| N. per confezione | 1 |
| Sterilità | Non-sterile |
| Vedi altri risultati |
† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.