Learn More
Thermo Scientific™ Nalgene™ Rectangular Polypropylene Tank with Cover
Codice prodotto. p-4525589
Click to view available options
Capacità (sistema metrico):
8 L
20 L
30 L
38 l
50 L
120 L
Dimensione della confezione:
Pezzo
Codice prodotto. 10232611
Marca: Thermo Scientific™ 142000002
Solo null disponibili
Aggiungi al carrello
Aggiungi al carrello
Significantly reduce evaporation and contamination by using these covered, rectangular tanks, which are offered in sizes from 8 to 114L to meet application needs.
Significantly reduce evaporation and contamination by using a Thermo Scientific™ Nalgene™ Rectangular Polypropylene Tank with Cover, offered in sizes ranging from 8 to 114L. Tanks can be modified with a factory-installed spigot.
  • Suitable for use with many organic chemicals
  • Offers good stress-crack resistance
  • Autoclavable
Note:
Tanks used at elevated temperatures or with high specific-gravity liquids should be externally supported.
TRUSTED_SUSTAINABILITY
Specifica
Lunghezza (sistema imperiale) 8 in.
Tipo Tank with Cover
Materiale Polypropylene
Capacità (sistema imperiale) 2 gal.
Capacità (sistema metrico) 8 L
Lunghezza (sistema metrico) 20 cm
Larghezza (sistema imperiale) 8 in.
Larghezza (sistema metrico) 20 cm
Altezza (sistema imperiale) 8 in.
Altezza (sistema metrico) 20 cm
Disclaimer to internal research use restriction:
Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Caution:
Plastic tanks are generally subject to more severe conditions that plastic labware; exposure is constant, stresses are greater and different classes and concentrations of chemicals are involved. Please pay special attention to chemical compatibility.