Learn More
Thermo Scientific™ Nalgene™ Storm Water Samplers and Mounting Kit
Descrizione
The Nalgene™ Storm Water Sampler is a convenient and affordable device for collecting quarterly storm water grab samples in compliance with EPA sampling requirements. No more standing in the rain waiting for water to flow or missed sampling events. The Storm Water Sampler can collect a full one liter grab sample of first flush storm water runoff through a storm water ditch, stream or storm grate outfall. Simply position the sampler in its protective mounting tube prior to a rain event, and leave it in place until after the storm.
- Leakproof† when polypropylene closure is applied
- Disposable sampling device is intended for one-time use
- Ideal for collecting samples for inorganic analyses
Includes:
EPDM gasket, polypropylene ball valve and closure
Specifica
Specifica
| Product Type | Water Sampler |
| Materiale | HDPE |
| Descrizione | Storm Water Sampler 1L HDPE |
| Include | HDPE Bottle |
| Materiale chiusura | Polypropylene |
| Capacità (sistema imperiale) | 33.8 oz. |
| Capacità (sistema metrico) | 1 L |
| Dimensione chiusura | 63 mm |
| Certificazione/Conformità | Meets EPA NPDES MSGP (Fed. Reg. FRL-6880-5 10/20/00 and most state grab sampling requirements |
| Quantità | 1/Pk., 4/Cs. |