missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nalgene™ Translucent Amber HDPE Diagnostic Bottles with Closure: Bulk Pack

Codice prodotto. p-4521830
Click to view available options
Capacità (sistema metrico):
4 mL
8 mL
15 mL
Finitura collo:
13-415 mm
20-415 mm
Dimensione della confezione:
pezzi 2000
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

Codice prodotto. 30117594

Marca: Thermo Scientific™ 3120849125

Solo null disponibili
Aggiungi al carrello
Aggiungi al carrello
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

Protect light-sensitive samples with translucent amber HDPE diagnostic bottles, intended for storage and shipping of small-volume, light-sensitive, valuable reagents.

Protect light-sensitive samples with Thermo Scientific™ Nalgene™ Translucent Amber HDPE Diagnostic Bottles with Closure, suitable for storage and shipping of small-volume, light-sensitive, valuable reagents. Translucent bottles provide light resistance with the ability to see liquid level and are ideal for test kits and many small-volume applications.

Features:
  • Nonsterile, high-density polyethylene bottles
  • Opaque, linerless polypropylene closures
  • Durable, uniform walls that resist splitting and puncturing
  • Non-cytotoxic
  • Leakproof†

Ordering Information:

Bottles and closures are packed in separate polybags within the same carton; 2,000 per case.

TRUSTED_SUSTAINABILITY

Specifica

Capacità 0.125 oz., 4 mL
Capacità (sistema metrico) 4 mL
Capacità (sistema imperiale) 0.125 oz.
Colore Translucent Amber
Descrizione 3.4mL, 20-415 closure, bulk pack
Diametro (sistema metrico) interno collo 8 mm
Materiale HDPE
N. per custodia 2000
N. per confezione 2000
Sterilità Non-sterile
Finitura collo 13-415 mm
Certificazione/Conformità Manufactured in compliance with ISO 13485:2016 quality requirements. Bottles and Closures meet the requirements of 21CFR177.1520 for food and beverage use, USP Class VI. Product meets the requirements for light-resistant containers per USP latest edition.
Closure Included Yes
Graduated No
Bocca Narrow
Forma Round
Altezza (sistema metrico) 41 mm
Product Type Sample Container
Larghezza (sistema imperiale) 0.62 in.
Altezza (sistema imperiale) 1.58 in.
Rivestimento interno chiusura Unlined
Materiale chiusura Polypropylene
Dimensione chiusura 13-415 mm
Diametro (sistema metrico) 16 mm
Tipo di chiusura Screw Cap
Erogazione attraverso la chiusura No
Vedi altri risultati Mostra meno risultati

† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.

Correzione del contenuto del prodotto

Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.

Titolo del prodotto

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.

Grazie per averci aiutato a migliorare il nostro sito web. Il vostro feedback è stato inviato