Learn More
Thermo Scientific™ Nalgene™ Utility Funnels
GREENER_CHOICE
Codice prodotto. p-4525020
Click to view available options
Diametro (sistema metrico) parte superiore:
65 mm
103 mm
155 mm
Capacità (sistema metrico):
62 ml
270 ml
1059 ml
Dimensione della confezione:
pezzi 12
Codice prodotto. 10365701
Marca: Thermo Scientific™ 42560234
Solo null disponibili
Aggiungi al carrello
Edge
Aggiungi al carrello
Replace glass lab funnels with these utility funnels, which are an excellent shatter-proof alternative.
Replace glass lab funnels with Thermo Scientific™ Nalgene™ Utility Funnels. These funnels are chemically resistant to a wide variety of lab chemicals and reagents and are shatterproof making them a valuble asset to lab safety programs.
  • 4256-0234 and 4256-0414 are made from shatter-proof, autoclavable polypropylene.
  • 4256-0638 has a large throat opening for faster transfer of materials and is made from shatter-proof high-density polyethylene (Not autoclavable).
TRUSTED_SUSTAINABILITY
Specifica
Diametro (sistema metrico) parte superiore 65 mm
Capacità (sistema metrico) 62 ml
Diametro (sistema metrico) gambo 10 mm
Materiale Polipropilene
Altezza (sistema metrico) 65 mm
Autoclavabile Autoclavable
Descrizione Funnel Utility PP 70mm
Lunghezza (sistema metrico) gambo 21 mm
Quantità 12/Pk.; 288/Cs.