Learn More
Nano-Secondary™ Alpaca, anti-Human, CoraLite Plus 488, Chromotek
Alpaca Recombinant Monoclonal Antibody
Marca: Chromotek shuGCL488-2-100
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
Nano-Secondary™ alpaca anti-human IgG, recombinant VHH, is a human IgG-specific (IgG1, IgG2, IgG3, IgG4) antibody. This secondary antibody product consists of a Nanobody that binds to the Fc-domain of human IgG with high affinity and specificity.
Specifica
| Nano-Secondary™ | |
| Alpaca | |
| Secondary | |
| Liquid |
| CoraLite Plus 488 | |
| 100 μL | |
| Human | |
| IgG |