Learn More
Thermo Scientific™ National Screw Thread Caps with Bonded Septa
Click to view available options
Pre-intagliato:
No
Yes
Dimensione chiusura:
10-425
13-425
8-425
Dimensione della confezione:
pezzi 100
Descrizione
Effectively eliminate septa push-through with Thermo Scientific™ National Screw Thread Caps with Bonded Septa.
- Septa and polypropylene caps bonded together for an integral system
- Eliminates septa push-through
- Available with choice of GPI thread sizes and preslit septa for easier thin-gauge needle penetration
Specifica
Specifica
| Descrizione | Black; Bonded Preslit Red 0.005 in. PTFE/White Silicone Septa; GPI Thread Finish: 13-425 |
| Materiale | PTFE, Silicone, Polypropylene |
| Pre-intagliato | Yes |
| Setto | Bonded Red PTFE/White Silicone, 0.005 in. |
| Product Line | National |
| Dimensione chiusura | 13-425 |