Learn More
Thermo Scientific™ National Target™ Assembled Vial, Cap and Septum Kits
Click to view available options
Marking Spot:
No
Sì
Colore chiusura:
Black
Light Blue
Setto:
PTFE/Silicone, Pre-slit
Red PTFE/White Silicone
Colore:
Amber
Clear
Dimensione della confezione:
pezzi 100
Descrizione
Thermo Scientific™ National Target Assembled Vial Kits include vials with pre-attached caps and septa, all packaged in convenient trays or polybags.
- Assembled kits include 100 vials with pre-attached caps and septa
- Packaged in convenient vial trays with clear covers or in polybags
- Vial trays are stackable for easy storage
- Vial trays hold vials upright for easy filling and maintaining sample order
Specifica
Specifica
| Colore chiusura | Light Blue |
| Colore (guarnizione) | Red/White |
| Descrizione | Amber Glass; Light Blue Cap; Red PTFE/White Silicone septum |
| Material (Liner) | PTFE/Silicone |
| Colore | Amber |
| Materiale | Glass |
| Setto | Red PTFE/White Silicone |
| Marking Spot | No |