Learn More
Thermo Scientific™ National Target DP™ Screw Caps with Septa
Click to view available options
Colore:
Blue
Ivory, Red, Blue
Red, Black
Setto:
Blue PTFE/Silicone
Ivory PTFE/Red Rubber
PTFE/Silicone
PTFE/Silicone/PTFE
Dimensione della confezione:
pezzi 100
Descrizione
Thermo Scientific™ National Target DP Screw Caps with Septa are color coded to separate assays, standards or samples.
- Preassembled with septa
Specifica
Specifica
| Descrizione | Ivory PTFE/Red Rubber Septa; Blue |
| Colore | Ivory, Red, Blue |
| Materiale | PTFE, Rubber |
| Setto | Ivory PTFE/Red Rubber |
| Product Line | National Target DP |
| Dimensione chiusura | 9-425 |