Learn More
Thermo Scientific™ National Target™ Snap-It™ Kits
Click to view available options
Marking Spot:
No
Sì
Colore chiusura:
Clear
Colore:
Clear
Setto:
PTFE/Red Rubber
PTFE/Silicone, Pre-slit
Dimensione della confezione:
pezzi 100
Descrizione
Thermo Scientific™ National Target Snap-It Convenience Kits provide vials and Snap-it caps with prefitted septa in storage trays.
- 2mL,12 × 32mm vials
- Choice of vial and preassembled seals with septa
- Packaged in two compartment benchtop storage trays
- Include matched quantities of vials and Snap-It caps with prefitted septa
- All items remain dust-free
Specifica
Specifica
| Colore chiusura | Clear |
| Descrizione | Clear Glass I-D; Septum: PTFE/Red Rubber |
| Colore | Clear |
| Materiale | PTFE, Rubber |
| Volume (sistema metrico) | 2 mL |
| Setto | PTFE/Red Rubber |
| Materiale chiusura | PTFE, Rubber |
| Marking Spot | Sì |