Learn More
Thermo Scientific™ National Target™ Unassembled Vial, Cap and Septum Convenience Kits
Click to view available options
Marking Spot:
No
Sì
Setto:
PTFE
PTFE/Silicone
PTFE/Silicone, Pre-slit
PTFE/Silicone/PTFE
Colore:
Amber
Clear
Materiale:
Glass
Silanized Glass
Dimensione della confezione:
pezzi 100
Descrizione
Keep vials, caps and septa within easy reach and dust free with Thermo Scientific™ National Target Unassembled Convenience Kits.
- Two-compartment benchtop storage tray
- I-D™ vials feature write-on patch with graduations
- All items remain handy and dust-free
- Caps with septa are not attached to vials (packed in separate polybag)
Includes:
100 vials and 100 caps with pre-assembled septa
Specifica
Specifica
| Colore chiusura | Light Blue |
| Descrizione | Clear Glass; Light Blue Cap, Pre-Slit Blue PTFE/White Silicone Septum; 2mL |
| Colore | Clear |
| Materiale | Glass |
| Pre-intagliato | Yes |
| Volume (sistema metrico) | 2 mL |
| Setto | PTFE/Silicone, Pre-slit |
| Marking Spot | No |