Learn More
Thermo Scientific™ National Unassembled Target LoVial™ Wide-Opening Vial Kits
Click to view available options
Volume (sistema metrico):
2 mL
4 mL
Setto:
PTFE/RR w/C4011-630 Insert
PTFE/Red Rubber
PTFE/Silicone
Colore:
Amber
Clear
Stile collo:
Standard
Wide
Dimensione della confezione:
pezzi 100
pezzi 500
Descrizione
Keep all vials and compatible crimp seals handy and dust-free with Thermo Scientific™ National Unassembled Target LoVial Wide-Opening Vial Kits.
- Two-compartment benchtop storage tray
- Include matched quantities of vials and silver aluminum seals with prefitted septa
Specifica
Specifica
| Colore chiusura | Silver |
| Descrizione | Amber Glass I-D; PTFE/RR with C4011-630 insert |
| Colore | Amber |
| Materiale | PTFE |
| Volume (sistema metrico) | 4 mL |
| Setto | PTFE/RR w/C4011-630 Insert |
| Stile collo | Wide |
| Materiale chiusura | Aluminum |
| Marking Spot | Sì |