Learn More
Abeba™ Nature 4040 Shoes
Click to view available options
Size:
34
35
37
39
40
41
42
43
44
45
46
47
48
Dimensione della confezione:
1 paio
Descrizione
- CE EN ISO 20347:2012 OB A, FO SRC
- Smooth leather
- White
- Slip resistance special rubber outsole sole
- Leather coated cork footbed
- Sanitized treated
- ESD dissipative according to DIN EN 61340
- Heel strap non-foldable, adjustable
- Adjustable instep area
- Buckle fastening
- Resistant to oil and benzine
Specifica
Specifica
| Certificazione/Conformità | CE EN ISO 20347:2012 OB A, FO SRC |
| Colore | White |
| Materiale pianta del piede | Cork |
| Materiale | Leather |
| Size | 34 |
| Caratteristiche suola | Slip-resistant |
| Materiale suola | Rubber |
| Stile | 4040 |