Learn More
Abeba™ Nature 4085 Shoes
Click to view available options
Size:
34
35
36
37
38
39
40
41
42
43
44
45
46
47
48
Dimensione della confezione:
1 paio
Descrizione
- CE EN ISO 20347:2012 OB A, FO SRC
- Smooth leather
- Black
- Slip resistance special rubber outsole sole
- Leather coated cork footbed
- Sanitized treated
- ESD dissipative according to DIN EN 61340
- 2 adjustable instep straps
- velcro fastening
Specifica
Specifica
| Certificazione/Conformità | CE EN ISO 20347:2012 OB A, FO SRC |
| Colore | Black |
| Materiale pianta del piede | Cork |
| Materiale | Leather |
| Size | 36 |
| Caratteristiche suola | Slip-resistant |
| Materiale suola | Rubber |
| Stile | 4085 |