missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Copper, Naval Brass, ARMI∣MBH
Click to view available options
Quantità:
1 Ea.
Dimensione della confezione:
Pezzo
Description
Copper, Naval Brass, Ø42x17mm
Specifications
Specifications
| Materiale o nome chimico | Silicon/Antimony/Bismuth/Phosphorus/Iron/Manganese/Arsenic/Lead/Nickel/Tin/Zinc/Copper/Aluminium/Cobalt |
| CAS | 7440-50-8 |
| Quantità | 1 Ea. |
| Forma | Disc |
| Concentration or Composition (by Analyte or Components) | Silicon 0.004 %/Antimony 0.0057 %/Bismuth 0.0065 %/Phosphorus 0.0223 %/Iron 0.037 %/Manganese 0.051 %/Arsenic 0.161 %/Lead 0.504 %/Nickel 0.52 %/Tin 0.535 %/Zinc 29.76 %/Copper 68.35 %/Aluminium (0.0004) %/Cobalt (0.0006) % |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction