Learn More
B7-H6 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Invitrogen PA554991
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
Immunogen sequence: TPLNDNVTIF CNIFYSQPLN ITSMGITWFW KSLTFDKEVK VFEFFGDHQE AFRPGAIVSP WRLKSGDASL RLPGIQLEEA GEYRCEVVVT PLKAQGTVQL EVVASPASRL LLDQVGMKEN EDKYMCESSG FY Highest antigen sequence identity to the following orthologs: Mouse - 27%, Rat - 28%.
Triggers NCR3-dependent natural killer cell activation.
Specifica
| B7-H6 | |
| Polyclonal | |
| Unconjugated | |
| NCR3LG1 | |
| Natural cytotoxicity triggering receptor 3 ligand 1, B7 homolog 6, B7-H6, NCR3LG1, B7H6 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 374383 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry, Western Blot | |
| 0.3 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q68D85 | |
| NCR3LG1 | |
| Recombit protein corresponding to Human NCR3LG1 | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |