Learn More
HUBER Silicone Hose, Pressureless, S-180-NW8
Codice prodotto. 10679152
Click to view available options
Materiale:
Silicone Rubber
Dimensione della confezione:
Pezzo
Codice prodotto. 10679152
Marca: HUBER 6077
Solo null disponibili
Aggiungi al carrello
Aggiungi al carrello
Suitable hose for pressureless applications and cooling water. HUBER; Silicone Hose, Pressureless, S-180-NW8 is not suitable for thermofluids that contain silicone.
  • Specifically designed for use in pressureless applications and with cooling water.
  • Silicone construction.
  • Thermally resistant.
  • Not for use with thermofluids that contain silicone.
Specifica
Materiale Silicone Rubber
Overall Length (Metric) 1 m