Learn More
Nectin-1 (CD111) Mouse anti-Human, BUV737, Clone: CK41, BD Biosciences™
Mouse Monoclonal Antibody
Marca: BD Biosciences 749460
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
Nectin-1 (CD111) Monoclonal antibody specifically detects Nectin-1 (CD111) in Human samples. It is validated for Flow Cytometry
Specifica
| Nectin-1 (CD111) | |
| Monoclonal | |
| Brilliant Ultraviolet 737 | |
| Nectin1; PRR1; PVRL1; PVRR1; HveC; HIgR; CLPED1; ED4; SK-12; OFC7 | |
| Human Recombinant Protein | |
| RUO | |
| Human | |
| IgG1 κ |
| Flow Cytometry | |
| CK41 | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Mouse | |
| 50 μg | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon BUV737 under optimal conditions that minimize unconjugated dye and antibody. |