missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NEDD4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Marca: Invitrogen™ PA579725
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: human U-87MG whole cell, human MDA-MB-453 whole cell.
The E3 ubiquitin-protein ligase NEDD4 is a Ubl conjugation pathway protein. It contains one HECT (E6AP-type E3 ubiquitin-protein ligase) domain and four WW domains. It is involved in the pathway leading to the degradation of VEGFR-2/KDFR, independently of its ubiquitin-ligase activity. It acts as a potential proto-oncogene that negatively regulates PTEN via ubiquitination, a paradigm analogous to that of Mdm2 and p53. It is also reported to be involved in the budding of many viruses and is the regulator of epithelial Na+ channels, which lead to arterial hypertension. NEDD4 protein is localized to the sarcolemmal region of muscle fibers. Increase in NEDD4 protein expression leads to unloaded and denervated muscles as well as a decrease in Notch1 expression. Hence it acts as down regulator of Notch-1 gene expression.
Specifica
| NEDD4 | |
| Polyclonal | |
| Unconjugated | |
| NEDD4 | |
| AA959633; AL023035; AU019897; cell proliferation-inducing gene 53 protein; E3 ubiquitin-protein ligase NEDD4; E430025J12Rik; HECT-type E3 ubiquitin transferase NEDD4; Kiaa0093; mKIAA0093; NEDD4; NEDD-4; NEDD4 E3 ubiquitin protein ligase; Nedd4-1; Nedd4a; Neural precursor cell expressed developmentally down-regulated protein 4; neural precursor cell expressed, developmentally down-regulated 4; neural precursor cell expressed, developmentally down-regulated 4, E3 ubiquitin protein ligase; neural precursor cell expressed, developmentally down-regulated gene 4; neural precursor cell expressed, developmentally down-regulated gene 4a; neural precursor cell expressed, developmentally down-regulted gene 4; OTTHUMP00000217840; OTTHUMP00000217841; OTTHUMP00000217842; PIG53; receptor-potentiating factor 1; RPF1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 17999, 25489, 4734 | |
| -20°C | |
| Lyophilized |
| ELISA, Western Blot | |
| 500 μg/mL | |
| PBS with 4mg trehalose and 0.05mg sodium azide | |
| P46934, P46935, Q62940 | |
| NEDD4 | |
| E. coli-derived human NEDD4 recombinant protein (Position: N960-F1245). | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto