Learn More
Aalborg™ Needle Valve for Stainless Steel Flowmeter
Codice prodotto. 11788808
Click to view available options
Velocità di flusso dell'aria:
20000 ml/min.;;en;
60000 mL/min.
Portata flusso d'acqua:
1200 ml/min.;;en;
3500mL/min.
Dimensione della confezione:
Pezzo
Codice prodotto. 11788808
Marca: Aalborg™ VCMSV
Solo null disponibili
Aggiungi al carrello
Aggiungi al carrello
These versatile, rugged and reliable, valves are suitable for laboratory instrumentation, bench top or OEM flow control purposes. Aalborg™ Needle Valve for Stainless Steel Flowmeter is designed for controlling a broad range of flow rates of liquids and gases.
  • Available in three conveniently overlapping orifice-needle sizes.
  • These versatile, rugged and reliable, valves are suitable for laboratory instrumentation, bench top or OEM flow control purposes.
  • Valves are offered in straight (T) and 90 degree (L) flow patterns. All valves are supplied with 1/8in. FNPT inlet and outlet ports.
  • Valve cartridges are also interchangeable with built-in valves of Aalborg’s series of P, T, S, and G flow meter product line.
  • Bubble tight shutoff
Specifica
Da utilizzare con (apparecchiatura) Per flussometro gas/liquido 150 mm
Tipo Valvola standard
Velocità di flusso dell'aria 20000 ml/min.;;en;
Portata flusso d'acqua 1200 ml/min.;;en;
Temperatura operativa max (sistema metrico) 121°C