Learn More
Nesprin-3 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Marca: Novus Biologicals NBP2-93890-0.02ml
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
Nesprin-3 Polyclonal antibody specifically detects Nesprin-3 in Human, Mouse samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
Specifica
| Nesprin-3 | |
| Polyclonal | |
| Western Blot 1:500-1:2000, Immunocytochemistry/ Immunofluorescence 1:50 - 1:200 | |
| chromosome 14 open reading frame 49, FLJ25605, MGC75267, nesprin-3, NET53 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 740-920 of human SYNE3 (NP_689805.3). LRLLEESLLSLIRNWHLQRMEVDSGKKMVFTNNIPKSGFLINPMDPIPRHRRRANLLQEEEGSHEDFSQLLRNFGQWLQVENSKLVRIIAMRTSTAEDLRTRKSKLQELEARVPEGQHLFENLLRLGPARGTSDELEDLRYQWMLYKSKLKDSGHLLTQSSPGEPTGFQKTRRWRGLGSLF | |
| 0.02 mL | |
| Cell Biology, Cytoskeleton Markers, Signal Transduction | |
| 161176 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunofluorescence | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |